missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Rabbit anti-Sheep, HRP, Secondary Antibody, Proteintech

Product Code. 17660297 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
1 mL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17660297 1 mL 1mL
17670297 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17660297 Supplier Proteintech Supplier No. SA00001161ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit anti-Sheep Affinipure Secondary Antibody

Specifications

Antigen IgG(H+L)
Applications ELISA, Western Blot
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Rabbit
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Sheep
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.