missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Rhodamine |
| Host Species | Rabbit |
| Quantity | 200 μL |
| Primary or Secondary | Secondary |
| Target Species | Goat |
| Form | Liquid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?