missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Donkey anti-Mouse, CoraLite594, Secondary Antibody, Proteintech

Product Code. 17640407 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
500 μL
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17640407 500 μL 500µL
17630407 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17640407 Supplier Proteintech Supplier No. SA000137500UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey anti-Mouse Affinipure Secondary Antibody

Specifications

Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate CoraLite 594
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Donkey
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.