missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Goat, AP, Secondary Antibody, Proteintech
Donkey anti-Goat Affinipure Secondary Antibody
Brand: Proteintech SA00002-3-200UL
This item is not returnable.
View return policy
Specifications
| IgG(H+L) | |
| Alkaline Phosphatase (AP) | |
| Donkey | |
| 200 μL | |
| Goat | |
| Liquid |
| ELISA, Western Blot | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction