missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG F(ab)2 Goat anti-Mouse, HRP, Polyclonal, Seconday Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-61929H-0.5ML
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgG F(ab)2 | |
| Polyclonal | |
| HRP | |
| Phosphate Buffered Saline (PBS) containing 0.2% BSA. | |
| Mouse IgG-F(ab')2 Fragment | |
| 0.5 mL | |
| Secondary | |
| Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 0.5 mg/mL | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry 1:10-1:500 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Antiserum was solid phase adsorbed to ensure class specificity. Antiserum was cross adsorbed using human and rat immunosorbents to remove cross reactive antibodies. The antibody to mouse F(ab')2 was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to horseradish peroxidase (HRP). Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse F(ab')2. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to human and rat F(ab')2 was detected. This antibody may cross react with F(ab')2 from other species. | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction