missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG F(ab')2, Goat anti-Human, HRP, Secondary Antlibody, Abnova™
Goat Secondary Antibody
Brand: Abnova PAB10698.2mg
This item is not returnable.
View return policy
Specifications
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| 2 mg | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Lyophilized |
| HRP | |
| Lyophilized from 20 mM KH2PO4, 150 mM NaCl, pH 7.2 (0.01% gentamicLyophilized from sulfate) | |
| Human IgG F(ab')2 fragment | |
| RUO | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction