missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Antibody (SPM556), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34428PE
404.60 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
Specifications
| IgG | |
| Monoclonal | |
| PE | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| 75 kDa | |
| 0.1 mL | |
| Immunology | |
| 3500 | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPM556 | |
| Flow Cytometry | |
| IGHG1 | |
| Purified human Ig Gamma Chain | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction