Learn More
IgG Antibody (SPM556), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34428APC
Product Code. 18105735
  • £508.00 / 0.10mL
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Monoclonal
APC
Immunoglobulin G, ImmunoglobulinG
Mouse
75 kDa
0.1 mL
Immunology
3500
Human
Purified
Flow Cytometry
SPM556
Flow Cytometry
IGHG1
Purified human Ig Gamma Chain
Protein A or G purified
RUO
Primary
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Store at 4C in the dark.
IgG2a κ