missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFL3 Monoclonal Antibody (536825)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524236
This item is not returnable.
View return policy
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IGF-I, rhIGF-II, rhIGFL-1, rhIGFL-4, and recombinant mouse IGFL-3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
IGFL3 belongs to the insulin-like growth factor (IGF) family of signaling molecules that play critical roles in cellular energy metabolism and in growth and development, especially prenatal growth.
Specifications
| IGFL3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6UXB1 | |
| IGFL3 | |
| E. coli-derived recombinant human IGFL-3 Thr25-Pro125. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| 536825 | |
| Unconjugated | |
| IGFL3 | |
| Gm580; IGF; IGF like family member 3; Igfl; Igfl3; IGF-like family member 3; Insulin growth factor-like family member; insulin growth factor-like family member 3; Insulin like growth factor; mIgfl; Rprc483; UNQ483; UNQ483/PRO982 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 388555 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction