missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP7 Recombinant Rabbit Monoclonal Antibody (24GB4910)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557429
This item is not returnable.
View return policy
Description
IGFBP7 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
Specifications
| IGFBP7 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IGFBP7 | |
| AGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7v; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3490 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4910 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16270 | |
| IGFBP7 | |
| A synthesized peptide derived from human IGFBP7 (aa 137-150). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction