missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP6 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518929
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma.
Specifications
| IGFBP6 | |
| Polyclonal | |
| Unconjugated | |
| IGFBP6 | |
| IBP6; IBP-6; IGF binding protein 6; IGF-binding protein 6; IGFBP; IGFBP6; IGF-BP6; Igfbp-6; Insulin like growth factor binding protein; insulin like growth factor binding protein 6; insulin-like growth factor binding protein 6; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 6 PRECURSOR (IGFBP-6) (IBP-6) (IGF-BINDING PROTEIN 6); insulin-like growth factor-binding protein 6; RBP6A | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3489 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P24592 | |
| IGFBP6 | |
| Synthetic peptide sequence (RPQDVNRRDQQRNP) corresponding to the internal amino acids of IGFBP6 (aa 128-142). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction