missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGFBP4 Monoclonal Antibody (A4), Invitrogen™

Mouse Monoclonal Antibody
£115.00 - £386.00
Specifications
| Antigen | IGFBP4 |
|---|---|
| Clone | A4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17654206
|
Invitrogen™
MA543751 |
20 μL |
£115.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17644206
![]() |
Invitrogen™
MA543750 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
| IGFBP4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| AI875747; Band 3A; BP-4; Deb2; differentially expressed in B16F10 2; HT29-IGFBP; IBP4; IBP-4; IGF-binding protein 4; IGFBP; Igfbp4; IGF-BP4; IGFBP-4; Insulin like growth factor binding protein; insulin like growth factor binding protein 4; insulin-like growth factor binding protein 4; insulin-like growth factor binding protein-4; insulin-like growth factor-binding protein 4; insulin-like growth factor-binding protein-4; LOW QUALITY PROTEIN: insulin-like growth factor-binding protein 4 | |
| IGFBP4 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| A4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P21744, P22692 | |
| 3487, 360622 | |
| Asp22-Glu258 (Accession P22692), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IGFBP4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title