Learn More
IGFBP3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30593-1-AP-20UL
Description
The IGFBP3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IGFBP3. This antibody recognizes human, mouse antigen. The IGFBP3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| IGFBP3 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| AI649005, IGFBP 3, Igfbp3, IGgfbp3 | |
| 20 μL | |
| 16009 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |