missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13981-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.Specifications
| IGFBP1 | |
| Polyclonal | |
| Unconjugated | |
| IGFBP1 | |
| AFBP, hIGFBP 1, IBP 1, IBP1, IGF binding protein 1, IGF BP25, IGFBP 1, IGFBP1, Placental protein 12, PP12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3484 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08833 | |
| IGFBP1 | |
| IGFBP1 Fusion Protein Ag5026 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction