missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-rP10/KAZALD1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98914V
This item is not returnable.
View return policy
Description
IGFBP-rP10/KAZALD1 Polyclonal antibody specifically detects IGFBP-rP10/KAZALD1 in Human samples. It is validated for Western Blot
Specifications
| IGFBP-rP10/KAZALD1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 81621 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| bone- and odontoblast-expressed gene 1, BONO1, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBP-rP10, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IGFBP-rP10/KAZALD1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction