missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-rp1/IGFBP-7 Rabbit anti-Human, Mouse, Rat, Clone: 8M10Q9, Novus Biologicals™

Product Code. 18321094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321094 100 μg 100µL
18394845 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321094 Supplier Novus Biologicals Supplier No. NBP316449100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGFBP-rp1/IGFBP-7 Monoclonal antibody specifically detects IGFBP-rp1/IGFBP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-rp1/IGFBP-7
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 8M10Q9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human IGFBP-rp1/IGFBP-7 (Q16270). LGETRDACGCCPMCARGEGEPCGGGGAGRGYCAPGMECVKSRKRRKGKAGAAAGGPGVSGVCVCKSRYPVCGSDGTTYPSGCQLRAASQRAESRGEKAITQ
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3490
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.