missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49861
This item is not returnable.
View return policy
Description
IGFBP-rp1/IGFBP-7 Polyclonal specifically detects IGFBP-rp1/IGFBP-7 in Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| IGFBP-rp1/IGFBP-7 | |
| Polyclonal | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_001544.1 | |
| IGFBP7 | |
| Peptide with sequence EKAITQVSKGTCEQG corresponding to internal region according to NP_001544.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. Cross reacts with Mouse. Expected from sequence similarity: Human and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3490 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction