missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody (115), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IGFBP-rp1/IGFBP-7 Monoclonal antibody specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IGFBP-rp1/IGFBP-7 |
| Clone | 115 |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-rp1/IGFBP-7 (Uniprot#: Q16270; Met1-Leu282). |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 3490 |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?