missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-rp1/IGFBP-7 Antibody (115), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90176AF594
This item is not returnable.
View return policy
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IGFBP-rp1/IGFBP-7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse IGFBP6 Human IGFBP4 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 115 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-rp1/IGFBP-7 (Q16270; Met1-Leu282). | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 3490 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction