missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-6 Antibody (001), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18362010 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362010 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362010 Supplier Novus Biologicals Supplier No. NBP290157AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGFBP-6 Monoclonal specifically detects IGFBP-6 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-6
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 750
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-6 (P24592; Met1-Gly240).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3489
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.