missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-5 Antibody (RM0097-12C23), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IGFBP-5 Monoclonal antibody specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IGFBP-5 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Quantity | 0.1 mL |
| Gene ID (Entrez) | 3488 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?