missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-5 Antibody (RM0097-12C23), DyLight 594, Novus Biologicals™

Product Code. 18422543 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422543 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422543 Supplier Novus Biologicals Supplier No. NBP122453DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IGFBP-5 Monoclonal antibody specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0097-12C23
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5
Host Species Rat
Immunogen Mouse recombinant protein of IGFBP-5
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3488
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.