missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-3 Rabbit anti-Human, Clone: 8, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen IGFBP-3
Clone 8
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18264716
View Documents
Novus Biologicals
NBP2-89495-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18299047
View Documents
Novus Biologicals
NBP2-89495-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications

Specifications

IGFBP-3
ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Monoclonal
Purified
RUO
Human
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Recombinant Human IGFBP-3 Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
8
ELISA
Unconjugated
Rabbit
Apoptosis, Biologically Active Proteins, Prostate Cancer
0.2 um filtered solution in PBS
3486
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.