Learn More
IGF2BP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14642-1-AP-20UL
 View more versions of this product
Product Code. 16863384
  • £135.00 / 20µL
Description
Both isoforms (64 kDa and 22 kDa) of IGFBP3 can be recognized by this antibody. IGFBP3 is nuclear and cytoplasmic in staining.
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some maligcies. IMP-3 is expressed in maligt melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of maligt melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
Specifications
Specifications
IGF2BP3
Polyclonal
Unconjugated
IGF2BP3
hKOC, IGF II mRNA binding protein 3, IGF2 mRNA binding protein 3, IGF2BP3, IMP 3, IMP3, KOC1, VICKZ family member 3, VICKZ3
Rabbit
Antigen Affinity Chromatography
RUO
10643, 140488
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00425, Q9CPN8
IGF2BP3
IGF2BP3 Fusion Protein Ag6226
20 μL
Primary
Mouse, Human