Learn More
IGF2BP2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11601-1-AP-20UL
Description
The IGF2BP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGF2BP2. This antibody recognizes human, mouse, rat antigen. The IGF2BP2 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| IGF2BP2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 10644 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP) | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |