missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGF2BP1 Recombinant Rabbit Monoclonal Antibody (K01_1S71)

Product Code. 30825012
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825012 50 μL
30826392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825012 Supplier Invitrogen™ Supplier No. MA565235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

IGF2BP1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

IGF2BP1 is a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2BP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1S71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene IGF2BP1
Gene Accession No. O88477, Q9NZI8
Gene Alias AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1
Gene Symbols IGF2BP1
Host Species Rabbit
Immunogen Recombinant protein of human IGF2BP1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10642, 140486
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.