Learn More
IGF2BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22803-1-AP-150UL
 View more versions of this product
Product Code. 16892095
  • £349.00 / 150µL
Description
MAST3, also named as KIAA0561, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. MAST3 catalyze the reaction: ATP + a protein = ADP + a phosphoprotein.
Specifications
Specifications
IGF2BP1
Polyclonal
Unconjugated
IGF2BP1
CRD BP, CRDBP, IGF II mRNA binding protein 1, IGF2 mRNA binding protein 1, IGF2BP1, IMP 1, IMP1, VICKZ family member 1, VICKZ1, ZBP 1, ZBP1, Zip code binding protein 1, Zipcode binding protein 1
Rabbit
Antigen Affinity Chromatography
RUO
10642, 140486, 303477
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88477, Q8CGX0, Q9NZI8
IGF2BP1
IGF2BP1 Fusion Protein Ag18859
150 μL
Primary
Human, Mouse, Rat