missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22803-1-AP-20UL
This item is not returnable.
View return policy
Description
MAST3, also named as KIAA0561, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. MAST3 catalyze the reaction: ATP + a protein = ADP + a phosphoprotein.Specifications
| IGF2BP1 | |
| Polyclonal | |
| Unconjugated | |
| IGF2BP1 | |
| CRD BP, CRDBP, IGF II mRNA binding protein 1, IGF2 mRNA binding protein 1, IGF2BP1, IMP 1, IMP1, VICKZ family member 1, VICKZ1, ZBP 1, ZBP1, Zip code binding protein 1, Zipcode binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10642, 140486, 303477 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88477, Q8CGX0, Q9NZI8 | |
| IGF2BP1 | |
| IGF2BP1 Fusion Protein Ag18859 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction