Learn More
Invitrogen™ IGF2BP1 Polyclonal Antibody

Description
Positive Samples: Jurkat, K-562, HepG2, HeLa, NIH/3T3, A-549, HL-60, Mouse liver; Cellular Location: Cell projection, Cytoplasm, Nucleus, axon, dendrite, dendritic spine, filopodium, growth cone, lamellipodium, perinuclear region Immunogen sequence: NFFGPKEEVK LETHIRVPAS AAGRVIGKGG KTVNELQNLT AAEVVVPRDQ TPDENDQVIV KIIGHFYASQ MAQRKIRDIL AQVKQQHQKG QSNQAQARRK.
Specifications
Specifications
| Antigen | IGF2BP1 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.59 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IGF2BP1 |
| Gene Accession No. | O88477, Q8CGX0, Q9NZI8 |
| Gene Alias | AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1 |
| Gene Symbols | IGF2BP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.