missing translation for 'onlineSavingsMsg'
Learn More

IGF-II R/IGF2R Rabbit anti-Human, Mouse, Rat, Clone: 4J4A3, Novus Biologicals™

Product Code. 18332794 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332794 20 μg 20µL
18345523 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332794 Supplier Novus Biologicals Supplier No. NBP31627020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGF-II R/IGF2R Monoclonal antibody specifically detects IGF-II R/IGF2R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-II R/IGF2R
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4J4A3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2392-2491 of human IGF-II R/IGF2R (P11717).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3482
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.