missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-II R/IGF2R Antibody (MEM-238), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IGF-II R/IGF2R Monoclonal antibody specifically detects IGF-II R/IGF2R in Human, Primate samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IGF-II R/IGF2R |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-238 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR |
| Host Species | Mouse |
| Immunogen | Recombinant Vaccinia virus encoding CD222. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?