missing translation for 'onlineSavingsMsg'
Learn More

IGF-II R/IGF2R Antibody (372604) [Allophycocyanin], Novus Biologicals™

Product Code. 30733353 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733353 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733353 Supplier Novus Biologicals Supplier No. FAB2447A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGF-II R/IGF2R Monoclonal antibody specifically detects IGF-II R/IGF2R in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-II R/IGF2R
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 372604
Conjugate APC
Formulation PBS
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGF-II R/IGF2R, Ser1510-Phe2108, Accession # P11717.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3482
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.