missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-II R/IGF2R Antibody (2G11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-514AF647
This item is not returnable.
View return policy
Description
IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Does not react with Chinese hamster ovary cell Mannose 6 Phosphate Receptor. | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2G11 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| IGF2R | |
| Purified bovine Mannose 6 Phosphate Receptor (Cation independent) [UniProt# P08169] | |
| 0.1 mL | |
| Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity | |
| 3482 | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction