missing translation for 'onlineSavingsMsg'
Learn More

IGF-II/IGF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£240.00 - £451.00

Specifications

Antigen IGF-II/IGF2
Dilution Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18677838
View Documents
Novus Biologicals
NBP2-48510-25ul
25 μL
£240.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18665397
View Documents
Novus Biologicals
NBP2-48510
0.1 mL
£451.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IGF-II/IGF2 Polyclonal antibody specifically detects IGF-II/IGF2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

IGF-II/IGF2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication, Cytokine Research
PBS (pH 7.2), 40% Glycerol
3481
IgG
Immunogen affinity purified
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Polyclonal
Purified
RUO
Human
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A
This antibody was developed against a recombinant protein corresponding to amino acids: TLQFVCGDRGFYFSRPASRVSRRSRGIVEECCFRSCDLALLETYCATPAKSERDVSTPPTVLPDNFPRYPVGKFFQYDTWKQSTQRLRRGLPALLRARRGHVLAKELEAFREAKRHRPLIALPTQD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.