missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IGF-II/IGF2 Antibody (DX-2647), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27979-100ug
This item is not returnable.
View return policy
Description
IGF-II/IGF2 Monoclonal antibody specifically detects IGF-II/IGF2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IGF-II/IGF2 | |
| Monoclonal | |
| Unconjugated | |
| P01344 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| DX-2647 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A | |
| IGF-2 | |
| 100 μg | |
| Cell Cycle and Replication, Cytokine Research | |
| 3481 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction