Learn More
Novus Biologicals™ IGF-II/IGF2 Antibody (DX-2647), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27979-50ug
 View more versions of this product
Product Code. 30044814
  • £162.00 / 50µg
Description
IGF-II/IGF2 Monoclonal antibody specifically detects IGF-II/IGF2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGF-II/IGF2
Monoclonal
Unconjugated
P01344
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
DX-2647
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A
IGF-2
50 μg
Cell Cycle and Replication, Cytokine Research
3481
Human
Purified