missing translation for 'onlineSavingsMsg'
Learn More

IGF-II/IGF2 Antibody (75015) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30736673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736673 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736673 Supplier Novus Biologicals Supplier No. FAB292JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGF-II/IGF2 Monoclonal antibody specifically detects IGF-II/IGF2 in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-II/IGF2
Applications Neutralization
Classification Monoclonal
Clone 75015
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A
Host Species Mouse
Immunogen E. coli-derived recombinant human IGF-II/IGF2, Ala25-Glu91, Accession # P01344.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3481
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.