missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IGF-II/IGF2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 110 |
| Conjugate | DyLight 405 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IGF-II/IGF2 (P09535; Ala25-Glu91). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?