missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IGF-I R/IGF1R Antibody (teprotumumab), Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Spezifikation
| Antigen | IGF-I R/IGF1R |
|---|---|
| Clone | teprotumumab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30046725
|
Novus Biologicals™
NBP3-28288-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044182
|
Novus Biologicals™
NBP3-28288-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044093
|
Novus Biologicals™
NBP3-28288-100ug |
100 μg |
£266.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpezifikation
| IGF-I R/IGF1R | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| IGF1R/ CD221 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| teprotumumab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P08069 | |
| 3480 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts