missing translation for 'onlineSavingsMsg'
Learn More

IGF-I/IGF-1 Rabbit anti-Human, Mouse, Rat, Rabbit, Clone: IGF1/2872R, Novus Biologicals™

Product Code. 18342473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342473 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342473 Supplier Novus Biologicals Supplier No. NBP30738120UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-I/IGF-1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone IGF1/2872R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Host Species Rabbit
Immunogen Recombinant full-length human IGF-I/IGF-1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3479
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.