missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I/IGF-1 Rabbit anti-Human, Mouse, Rat, Rabbit, Clone: IGF1/2872R, Novus Biologicals™
Rabbit Monoclonal Antibody
£440.00
Specifications
| Antigen | IGF-I/IGF-1 |
|---|---|
| Clone | IGF1/2872R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| IGF-I/IGF-1 | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 3479 | |
| IgG | |
| Protein A or G purified |
| IGF1/2872R | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Rabbit | |
| IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
| Recombinant full-length human IGF-I/IGF-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title