missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals IGF-I/IGF-1 Antibody (SPM406) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34409-0.1mg
This item is not returnable.
View return policy
Description
IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| IGF-I/IGF-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5-1 μg/mL, Flow Cytometry 0.5-1 μg/million cells, ELISA 1-5μg/mL for coating, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, CyTOF-ready | |
| IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
| Purified human IGF-I/IGF-1 protein (Uniprot: P05019) | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 3479 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| SPM406 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction