missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I/IGF-1 Antibody (IGF1/1020), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48001JF646
This item is not returnable.
View return policy
Description
IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOF
Specifications
| IGF-I/IGF-1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
| Recombinant full-length human IGF-I/IGF-1 protein (Uniprot: P05019) | |
| 0.1 mL | |
| BiologicAll speciesy Active Proteins, Cell Cycle and Replication, Signal Transduction | |
| 3479 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| IGF1/1020 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction