missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I/IGF-1 Antibody (IGF1/1020), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
£463.00 - £501.00
Specifications
| Antigen | IGF-I/IGF-1 |
|---|---|
| Clone | IGF1/1020 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636748
|
Novus Biologicals
NBP2-48001B-100ul |
100 μg |
£501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656328
|
Novus Biologicals
NBP2-48001B |
0.1 mL |
£463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| IGF-I/IGF-1 | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Cell Cycle and Replication | |
| PBS | |
| 3479 | |
| IgG1 κ | |
| Protein A or G purified |
| IGF1/1020 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Rabbit | |
| IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
| Recombinant full-length human IGF-I/IGF-1 protein (Uniprot: P05019) | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title