missing translation for 'onlineSavingsMsg'
Learn More

IGF-I/IGF-1 Antibody (56408) [HRP], Novus Biologicals™

Product Code. 30734184 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734184 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734184 Supplier Novus Biologicals Supplier No. FAB291H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-I/IGF-1
Applications Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 56408
Conjugate HRP
Formulation PBS
Gene Alias IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Host Species Mouse
Immunogen E. coli-derived recombinant human IGF-I/IGF-1, Gly49-Ala118, Accession # P01343
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3479
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.