missing translation for 'onlineSavingsMsg'
Learn More

Igepal CO-520, TRC

Product Code. 30399345
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399345 10 g 10g
30389903 1 g -
30394775 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399345 Supplier Toronto Research Chemicals Supplier No. TRCI26505010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68412-54-4
Molecular Formula (C2H4O)n.C15H24O
Synonym ?-(Nonylphenyl)-?-hydroxy-poly(oxy-1,2-ethanediyl) Branched Polymer, Manual Registration, Generic Registration

Specifications

Quantity 10 g
Chemical Name or Material 4-Nonylphenol-ethoxylate (penta-) (technical) (branched)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.