missing translation for 'onlineSavingsMsg'
Learn More

IGEPAL CA-630, TRC

Product Code. 30389075
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389075 25 g
30402923 50 g
30394915 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389075 Supplier Toronto Research Chemicals Supplier No. TRCI94202325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9036-19-5
Molecular Formula (C2H4O)nC14H22O
Synonym α-[(1,1,3,3-Tetramethylbutyl)phenyl]-ω-hydroxy-poly(oxy-1,2-ethanediyl), Polyethylene glycols mono[(1,1,3,3-tetramethylbutyl)phenyl] ether, APEO 12, Alkasurf OP, Alkasurf OP 10, Alkasurf OP 12, Alkasurf OP 30, Alkasurf OP 40, Alkasurf OP 5, Alkasurf OP 8, Blaunon NK 8055, Blaunon NK 808, CA 897, Cemulsol OP 16, Cemulsol P 9, Charger E, Companion, Companion (surfactant), DeIonic OPE 10, Disponil A 4065EXP, EA 142, EA 142B, EP 680, Emulgen 808, Emulgen 810, Emulgen 810S, Emulgen 840S, Emulsifier OP, Ethoxylated octylphenol, Ethylan CP, Ethylan CPX, HS 2045, HS 208, HS 210, HS 215, Hydrol, Hydrol (surfactant), Hyonic OP 9, Hyonic PE 260, Igepal 603, Igepal CA, Igepal CA 210, Igepal CA 300, Igepal CA 360, Igepal CA 420, Igepal CA 520, Igepal CA 620, Igepal CA 630, Igepal CA 720, Igepal CA 877, Igepal CA 887, Igepal CA 890, Igepal CA 897, Invadine JFC 800, Macol OP 10, Macol OP 10SP, Macol OP 5, Makon OP 9, Marlophen 84, Marlophen 85, Monopol OP 1013, Monopol OP 1015, Monopol OP 1019, NK 8055, NP 40, Nekanil LN, Neutronyx 622, Neutronyx 675, Newcol 804, Newcol 808, Newcol 860, Newcol 864, Newcol 865, Nikkol DP 3, Nikkol OP, Nikkol OP 10, Nikkol OP 18TX, Nikkol OP 5, Nissan HS 240, Nissan Nonion HS, Nissan Nonion HS 204.5, Nissan Nonion HS 206, Nissan Nonion HS 208, Nissan Nonion HS 210, Nissan Nonion HS 215, Nissan Nonion HS 220, Nissan Nonion HS 230, Nissan Nonion HS 240, Nissan Nonion HS 270, Noigen EA 102, Noigen EA 110, Noigen EA 112, Noigen EA 120, Noigen EA 140, Noigen EA 142, Noigen EA 142B, Noigen EA 151, Noigen EA 160, Noigen EA 170, Noigen EA 190D, Noigen EA 92, Nonal 109, Nonal 310, Nonidet P 40, Nonion HS 2045, Nonion HS 206, Nonion HS 208, Nonion HS 210, Nonion HS 220, Nonion HS 240, OF 7, OP, OP (Chinese surfactant), OP 1, OP 10, OP 1062, OP 115, OP 12, OP 13.2, OP 15, OP 17.7, OP 2, OP 2 (Chinese surfactant), OP 20, OP 20 (Chinese surfactant), OP 21, OP 21 (surfactant), OP 3, OP 3 (Chinese surfactant), OP 30, OP 4, OP 40, OP 6, OP 7, OPE 10, Octapol 100, Octapol 80, Octoxynol, Octoxynol 10, Octoxynol 11, Octoxynol 13, Octoxynol 3, Octoxynol 33, Octoxynol 40, Octoxynol 70, Octoxynol 9, Octylphenol ethoxylate, Octylphenoxypoly(ethoxyethanol), Octylphenoxypoly(ethyleneoxy)ethanol, Peregal OP 10, Phenoxol 8/10, Pionin D 310, Poly(ethylene oxide) octylphenyl ether, Poly(oxy-1,2-ethanediyl), α-hydro-ω-hydroxy- octylphenyl ether, Poly(oxyethylene) octylphenol ether, Poly(oxyethylene) octylphenyl ether, Polyethylene glycol (tetramethylbutyl)phenyl ether, Polyethylene glycol mono(octylphenyl) ether, Polyethylene glycol mono(tert-octylphenyl) ether, Polyethylene glycol monooctylphenol ether, Polyethylene glycol octylphenyl ether, Polyethylene glycol tert-octylphenyl ether, Polyoxyethylene monooctylphenyl ether, Polyoxyethylene octylphenyl ether, R 11, R 11 (surfactant), Renex 756, Rokafenol O, Rokafenol O 22, Secopal OP 20, Serdox NOP 9, Sinnopal OP, Sinnopal OP 8, Siponic F 160, Siponic F 300, Siponic F 400, Siponic F 707, Surfonic OP 10, Synperonic OP, Synperonic OP 10, Synperonic OP 10EO, Synperonic OP 11, Synperonic OP 11EO, Syntopon 8D1, Syntopon A 100, T 45, T 45 (polyglycol), T-DET O 407, TX 10, TX 10 (polyoxyalkylene), TX 114, TX 25, Teric GX 13, Teric X, Teric X 10, Teric X 11, Teric X 16, Teric X 40, Teric X 5, Teric X 8, Triton AG 98, Triton X 110, Triton X 144, Trixon 100, Value 3608, X 350, tert-Octylphenoxy poly(oxyethylene)ethanol, tert-Octylphenoxypoly(ethoxyethanol)
IUPAC Name 4-Octylphenol-ethoxylate
SMILES CC(C)(CC(C)(C1=CC=C(OCCO)C=C1)C)C

Specifications

Quantity 25 g
Chemical Name or Material 4-iso-Octylphenol-ethoxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.