missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgE, Biotin, clone: 23G3, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 13-5992-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The 23G3 monoclonal antibody reacts with the ε heavy chain of mouse IgE. It does not react with other classes of mouse immunoglobulin including IgD, IgG, IgA, or IgM.Applications Reported: The 23G3 antibody has been reported for use in flow cytometric analysis, and ELISA.Applications Tested: The 23G3 antibody has been tested by ELISA and can be used at 1:5000-1:10000. This 23G3 antibody has also been tested by flow cytometric analysis of IGEL cells, and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.Specifications
IgE | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Conjugated | |
Ighe | |
IgG1, kappa | |
100μg | |
Store at 2-8°C. Do not freeze. | |
Primary | |
380792 |
Flow Cytometry | |
23G3 | |
Biotin | |
Ighe | |
Immunoglobulin E, Ig E | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
For Research Use Only.