missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Mouse anti-Human, Clone: 4D6, Abnova™
Mouse monoclonal antibody raised against human IgE.
Brand: Abnova MAB12444.1mg
This item is not returnable.
View return policy
Specifications
| IgE | |
| Monoclonal | |
| Unconjugated | |
| ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Antigen Affinity Purified | |
| RUO | |
| This product is an IgG2a kappa antibody that was purified from tissue culture. It reacts with human IgE by indirect ELISA and does not react with IgM, IgA or IgG. > 90% pure as determined by page gel. | |
| Store at 4°C. After reconstitution with 1mL of reagent quality water, aliquots should be stored at −20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a |
| ELISA, Immunoprecipitation, Western Blot | |
| 4D6 | |
| Mouse monoclonal antibody raised against human IgE. | |
| Liquid | |
| Human IgE whole molecule. | |
| 1 mg | |
| Secondary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction