missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Monoclonal Antibody (Ige21), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen H077T03R04
This item is not returnable.
View return policy
Description
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
Specifications
| IgE | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide, pH 19.2 | |
| P01854 | |
| IGHE | |
| Affinity chromatography | |
| RUO | |
| 3497 | |
| 4°, C, store in dark, DO NOT FREEZE! | |
| IgG1 κ |
| Flow Cytometry | |
| Ige21 | |
| NovaFluor Red 710 | |
| IGHE | |
| HCAK1, IgA1, IgG3, IGHA1, IGHA2, IGHG1, IGHG2, IGHG3, IGHG4, IGKC, IGKCD, IGLC, IGLC1, Immunoglobulin E, immunoglobulin heavy constant alpha 1, immunoglobulin heavy constant alpha 2 (A2m marker), immunoglobulin heavy constant gamma 1 (G1m marker), immunoglobulin heavy constant gamma 2 (G2m marker), immunoglobulin heavy constant gamma 3 (G3m marker), immunoglobulin heavy constant gamma 4 (G4m marker), immunoglobulin kappa constant, immunoglobulin lambda constant 1, ImmunoglobulinE, Km | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction